missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX9 Antibody (CL0639) [Janelia Fluor™ 646], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52943JF646
This item is not returnable.
View return policy
Description
SOX9 Monoclonal antibody specifically detects SOX9 in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/ Immunofluorescence
Specifications
| SOX9 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| campomelic dysplasia, autosomal sex-reversal, CMD 1, CMD1, CMPD1, SRA1SRY (sex-determining region Y)-box 9 protein, SRY (sex determining region Y)-box 9, SRY-related HMG-box, gene 9, transcription factor SOX-9 | |
| This antibody was developed using a recombinant protein derived from P48436, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, Phospho Specific, Stem Cell Markers | |
| 6662 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown, Immunocytochemistry/Immunofluorescence | |
| CL0639 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction