Learn More
SOX9 Antibody (CL0639) [Alexa Fluor™ 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52943AF350
Product Code. 30303939
  • £582.00 / 0.10mL
Description
SOX9 Monoclonal antibody specifically detects SOX9 in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SOX9
Monoclonal
Alexa Fluor 350
campomelic dysplasia, autosomal sex-reversal, CMD 1, CMD1, CMPD1, SRA1SRY (sex-determining region Y)-box 9 protein, SRY (sex determining region Y)-box 9, SRY-related HMG-box, gene 9, transcription factor SOX-9
This antibody was developed using a recombinant protein derived from P48436, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Phospho Specific, Stem Cell Markers
6662
Store at 4⪚C in the dark.
IgG2a
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown, Immunocytochemistry/Immunofluorescence
CL0639
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human, Mouse
Purified