missing translation for 'onlineSavingsMsg'
Learn More

SOX9 Antibody (326820) [Janelia Fluor™ 646], Novus Biologicals™

Product Code. 30737501 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30737501 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737501 Supplier Novus Biologicals Supplier No. FAB3075J

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SOX9 Monoclonal antibody specifically detects SOX9 in Human samples. It is validated for Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOX9
Applications Immunocytochemistry
Classification Monoclonal
Clone 326820
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias campomelic dysplasia, autosomal sex-reversal, CMD 1, CMD1, CMPD1, SRA1SRY (sex-determining region Y)-box 9 protein, SRY (sex determining region Y)-box 9, SRY-related HMG-box, gene 9, transcription factor SOX-9
Host Species Mouse
Immunogen E. coli-derived recombinant human SOX9, Met1-Lys151, Accession # P48436
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6662
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.