missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX8 Antibody (5A7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00030812-M03
This item is not returnable.
View return policy
Description
SOX8 Monoclonal antibody specifically detects SOX8 in Human samples. It is validated for Western Blot, ELISA
Specifications
| SOX8 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| MGC24837, SRY (sex determining region Y)-box 8, transcription factor SOX-8 | |
| SOX8 (NP_055402, 102 a.a. ~ 210 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. VKRPMNAFMVWAQAARRKLADQYPHLHNAELSKTLGKLWRLLSESEKRPFVEEAERLRVQHKKDHPDYKYQPRRRKSAKAGHSDSDSGAELGPHPGGGAVYKAEAGLGD | |
| 0.05 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 5A7 | |
| Western Blot 1:500, ELISA | |
| NP_055402 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 30812 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction