missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ SOX7 Recombinant Protein Antigen

Código de producto. 18393660 Tienda Bio Techne Productos
Change view
Click to view available options
Quantity:
0.1mL
Tamaño de la unidad:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity unitSize
18393660 0.1mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18393660 Proveedor Novus Biologicals™ N.º de proveedor NBP186116PEP

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human SOX7. The SOX7 Recombinant Protein Antigen is derived from E. coli. The SOX7 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.

This is a blocking peptide for NBP1-86116. This is a human recombinant protein expressed in E. coli with a N-terminal His-ABP tag and purified with IMAC chromatography.

TRUSTED_SUSTAINABILITY

Especificaciones

Gene ID (Entrez) 83595
Species Human
Purification Method Chromatography
Purity >80%
Concentration 0.5mg/mL
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Formulation PBS and 1M Urea, pH 7.4.
For Use With (Application) Blocking/Neutralizing, Control
Gene Symbol SOX7
Label Type Unlabeled
Molecular Weight (g/mol) 32kDa
Product Type SOX7
Quantity 0.1mL
Regulatory Status RUO
Source E.Coli
Specific Reactivity This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP1-86116. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
Immunogen SPLHCSHPLGSLALGQSPGVSMMSPVPGCPPSPAYYSPATYHPLHSNLQAHLGQLSPPPEHPGFDALDQLSQVELLGDMDRNEFDQYLNTPGHPDSATGAMALSGHVPVSQVTPTGPTETSLISVLADATATYYNSYS
Mostrar más Mostrar menos

For Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.