Learn More
Invitrogen™ SOX7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA580055
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Brain Tissue, Human Placenta Tissue, Rat Lung Tissue, Rat Testis Tissue, HELA whole cell, A549 whole cell, HEPG2 whole cell, SMMC whole cell, NEURO whole cell.
This gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. The protein may play a role in tumorigenesis. A similar protein in mice is involved in the regulation of the wingless-type MMTV integration site family (Wnt) pathway.
Specifications
| SOX7 | |
| Polyclonal | |
| Unconjugated | |
| Sox7 | |
| MGC10895; mSOX7; SOX7; Sox-7; SRY (sex determining region Y)-box 7; SRY box 7; SRY-box 7; SRY-box containing gene 7; SRY-box transcription factor 7; transcription factor SOX-7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20680, 290317, 83595 | |
| -20°C | |
| Lyophilized |
| Western Blot | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
| P40646, Q9BT81 | |
| Sox7 | |
| A synthetic peptide corresponding to a sequence at the C-terminus of human SOX7 (370-388aa LISVLADATATYYNSYSVS). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.