missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SOX6 Monoclonal Antibody (CL5685)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531425
This item is not returnable.
View return policy
Description
Immunogen sequence: NLDTFEHGGG HSYNHKQIE Highest antigen sequence indentity to the following orthologs: Mouse - 84%, Rat - 89%.
The SOX6 gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors, which are known to play vital roles in the regulation of developmental processes and cell fate determination. SOX6, along with its close relatives SOX5 and SOX13, constitutes the SOXD subfamily, which is involved in the regulation of the development of the central nervous system, skeletal system, and cartilage. It functions by binding to specific DNA sequences and modulating the expression of target genes necessary for these developmental processes. SOX6 has been implicated in various functions, such as promoting cartilage formation by acting in conjunction with SOX9 to drive chondrogenesis. In adult neural stem cells, SOX6 is essential for the activation of radial glial-like cells and the generation of new neurons, emphasizing its role in both embryonic development and adult tissue maintenance. The dysregulation of SOX6 has been associated with developmental disorders and its potential involvement in pathologies like cancer, where it influences cell differentiation and proliferation pathways.
Specifications
| SOX6 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P35712 | |
| SOX6 | |
| Synthetic peptide corresponding to Human SOX6. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| CL5685 | |
| Unconjugated | |
| SOX6 | |
| AI987981; HSSOX6; SOX6; Sox-6; SOXD; SOX-LZ; SRY (sex determining region Y)-box 6; SRY box 6; SRY-box 6; SRY-box containing gene 6; transcription factor SOX-6 | |
| Mouse | |
| Protein A | |
| RUO | |
| 55553 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction