missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX6 Antibody (CL5690) [mFluor Violet 610 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61423MFV610
This item is not returnable.
View return policy
Description
SOX6 Monoclonal antibody specifically detects SOX6 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| SOX6 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| HSSOX6, SOXD, SRY (sex determining region Y)-box 6, SRY-box containing gene 6, transcription factor SOX-6 | |
| This antibody was developed using a synthetic peptide derived from P35712, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL5690 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 55553 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction