missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX6 Antibody (CL5685), Novus Biologicals™
Mouse Monoclonal Antibody
£328.00 - £470.00
Specifications
| Antigen | SOX6 |
|---|---|
| Clone | CL5685 |
| Dilution | Western Blot 1:500 - 1:1000, Immunohistochemistry-Paraffin |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18267261
|
Novus Biologicals
NBP2-61422 |
100 μL |
£470.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18656086
|
Novus Biologicals
NBP2-61422-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SOX6 Monoclonal specifically detects SOX6 in Human samples. It is validated for Western Blot.Specifications
| SOX6 | |
| Western Blot 1:500 - 1:1000, Immunohistochemistry-Paraffin | |
| Unconjugated | |
| Mouse | |
| HSSOX6, SOXD, SRY (sex determining region Y)-box 6, SRY-box containing gene 6, transcription factor SOX-6 | |
| SOX6 | |
| IgG1 | |
| Protein A purified |
| CL5685 | |
| Monoclonal | |
| Purified | |
| Human | |
| 55553 | |
| This antibody was developed against a synthetic peptide corresponding to amino acids: NLDTFEHGGGHSYNHKQIE | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title