missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13216-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. The encoded protein may play a role in chondrogenesis. A pseudogene of this gene is located on chromosome 8. Multiple transcript variants encoding distinct isoforms have been identified for this gene.Specifications
| SOX5 | |
| Polyclonal | |
| Unconjugated | |
| SOX5 | |
| L SOX5, SOX5, Transcription factor SOX 5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 140587, 20678, 6660 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P35710, P35711 | |
| SOX5 | |
| SOX5 Fusion Protein Ag4092 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction