missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX5 Antibody (OTI2A8), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-74301CL1
This item is not returnable.
View return policy
Description
SOX5 Monoclonal antibody specifically detects SOX5 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| SOX5 | |
| Monoclonal | |
| CoraFluor 1 | |
| L-SOX5, MGC35153, SRY (sex determining region Y)-box 5, transcription factor SOX-5 | |
| Full length human recombinant protein of human SOX5(NP_008871) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| OTI2A8 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 6660 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction