missing translation for 'onlineSavingsMsg'
Learn More

SOX4 Antibody (PCRP-SOX4-1D6), DyLight 550, Novus Biologicals™

Product Code. 18361883
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18361883 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18361883 Supplier Bio-Techne Supplier No. NBP314180R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SOX4 Monoclonal antibody specifically detects SOX4 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOX4
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PCRP-SOX4-1D6
Conjugate DyLight 550
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ecotropic viral integration site 16, EVI16, SRY (sex determining region Y)-box 4, SRY-related HMG-box gene 4, transcription factor SOX-4
Host Species Mouse
Immunogen Recombinant fragment (around aa45-130) of human SOX4 protein (Uniprot: Q06945)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 6659
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.