Learn More
Invitrogen™ SOX3 Monoclonal Antibody (CL4725)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524710
Description
Immunogen sequence: RENSSGARSPR VPADLARSIL ISLPFPPDSL AHRPPSSAPT ESQGLFTVAA PAPGAPSPPA TLAHLLPAPA MYSLLETELK NPVGTPTQAA GTGGPAAPGG AGKSSA Binds to an epitope located within the peptide sequence LISLPFPPDS as determined by overlapping synthetic peptides. Highest antigen sequence identity to the following orthologs - mouse 82%, rat 31%.
This gene encodes a member of the SOX family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. Mutations in this gene have been associated with X-linked mental retardation with growth hormone deficiency.
Specifications
| SOX3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P41225 | |
| SOX3 | |
| Recombinant protein fragment of Human SOX3 (Product #RP-108155) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| CL4725 | |
| Unconjugated | |
| SOX3 | |
| GHDX; LOC679158; MRGH; PHP; PHPX; similar to SRY (sex determining region Y)-box 3; Sox3; Sox-3; SOXB; SRY (sex determining region Y)-box 3; SRY box 3; SRY-box 3; SRY-box containing gene 3; transcription factor SOX-3 | |
| Mouse | |
| Protein A | |
| RUO | |
| 6658 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.