missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX21 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55052-1-AP-150UL
This item is not returnable.
View return policy
Description
The SOX gene family is defined by the Sry-related HMG box domain that mediates sequence-specific DNA binding. A variety of functions are associated with the SOX proteins during development. The highly complex group of Sox genes cluster at least 40 different loci that rapidly diverged in various animal lineages. At present, 30 Sox genes have been identified. Members of this family have been shown to be conserved during evolution and to play key roles during animal development. SOX21 may play a role as an activator of transcription of OPRM1. Overexpression of SOX21 can cause an up-regulation of the OPRM1 distal promoter in mor-expressing neuronal cells.Specifications
| SOX21 | |
| Polyclonal | |
| Unconjugated | |
| SOX21 | |
| SOX A, SOX21, SOX25, SOXA, Transcription factor SOX 21 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11166, 223227 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.53 mg/mL | |
| 0.1 M glycine, 0.1 M sodium bicarbonate with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q811W0, Q9Y651 | |
| Sox21 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction