missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SOX21 Monoclonal Antibody (CL4688)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531403
This item is not returnable.
View return policy
Description
Immunogen sequence: GKPQLDPYPA AYAAAL Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.
The SOX gene family is defined by the Sry-related HMG box domain that mediates sequence-specific DNA binding. A variety of functions are associated with the SOX proteins during development. The highly complex group of Sox genes cluster at least 40 different loci that rapidly diverged in various animal lineages. At present, 30 Sox genes have been identified. Members of this family have been shown to be conserved during evolution and to play key roles during animal development. SOX21 may play a role as an activator of transcription of OPRM1. Overexpression of SOX21 can cause an up-regulation of the OPRM1 distal promoter in mor-expressing neuronal cells.
Specifications
| SOX21 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q811W0, Q9Y651 | |
| Sox21 | |
| Synthetic peptide corresponding to Human SOX21. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| CL4688 | |
| Unconjugated | |
| Sox21 | |
| DNA-binding protein SOX21; HMG-box protein SOX21; Sox21; Sox25; SOX25SOX-A; SOXA; SOX-A; SRY (sex determining region Y)-box 21; SRY box 21; SRY-box 21; SRY-box containing gene 21; transcription factor SOX-21 | |
| Mouse | |
| Protein A | |
| RUO | |
| 11166, 223227 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction