missing translation for 'onlineSavingsMsg'
Learn More

SOX21 Antibody (OTI7D5) [mFluor Violet 450 SE], Novus Biologicals™

Product Code. 30687856 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30687856 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30687856 Supplier Novus Biologicals Supplier No. NBP344697MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SOX21 Monoclonal antibody specifically detects SOX21 in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOX21
Applications Western Blot
Classification Monoclonal
Clone OTI7D5
Conjugate mFluor Violet 450 SE
Formulation 50mM Sodium Borate
Gene Alias SOX25SOX-A, SOXA, SRY (sex determining region Y)-box 21, SRY-box 21, transcription factor SOX-21
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-140 of human SOX21(NP_009015) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11166
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.