missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX21 Antibody (CL4688) [PerCP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59059PCP
This item is not returnable.
View return policy
Description
SOX21 Monoclonal antibody specifically detects SOX21 in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| SOX21 | |
| Monoclonal | |
| PerCP | |
| SOX25SOX-A, SOXA, SRY (sex determining region Y)-box 21, SRY-box 21, transcription factor SOX-21 | |
| This antibody was developed using a synthetic peptide derived from Q9Y651, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL4688 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 11166 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction