Learn More
Invitrogen™ SOX2 Recombinant Rat Monoclonal Antibody (Btjce), HRP, Invitrogen™
Rat Recombinant Monoclonal Antibody
£171.00 - £343.00
Specifications
| Antigen | SOX2 |
|---|---|
| Clone | Btjce |
| Concentration | 1.0 mg/mL |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30130875
|
Invitrogen™
740013THRP20UG |
20 μL |
£171.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30132368
|
Invitrogen™
740013THRP |
50 μL |
£343.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Recombinant rat monoclonal antibodies are produced using in vitro expression systems. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include: better specificity, lot-to-lot consistency, and broader immunoreactivity to diverse targets due to a larger rabbit immune repertoire. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
SOX2 Recombinant Monoclonal antibody specifically detects SOX2 in Human samples. It is validated for Western BlotSpecifications
| SOX2 | |
| 1.0 mg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with proprietary stabilizer and proprietary preservative; pH 7.4 | |
| ANOP3; cb236; Delta EF2a; lcc; MCOPS3; MCOPS3 (Microphthalmia Syndromic type 3); RGD1565646; sex determining region Y-box 2; SOX 2; SOX2; Sox-2; soxp; SRY (sex determining region Y) box 2; SRY (sex determining region Y)-box 2; SRY box 2; SRY related HMG box 2; SRY-box 2; SRY-box 2; SOX2; SRY-box containing gene 2; SRY-box transcription factor 2; SRY-related HMG-box gene 2; transcription factor SOX2; transcription factor SOX-2; wu:fb83g04; wu:fc14d07; ysb; zgc:65860; zgc:77389 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
| Btjce | |
| Western Blot | |
| HRP | |
| Rat | |
| Human | |
| P48431 | |
| 6657 | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| SOX2 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.