missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SOX2 Recombinant Rat Monoclonal Antibody (Btjce), Alexa Fluor™ Plus 555, Invitrogen™
Rat Recombinant Monoclonal Antibody
Brand: Invitrogen™ 740013TP55520UG
This item is not returnable.
View return policy
Description
SOX2 Recombinant Monoclonal antibody specifically detects SOX2 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin)
SOX2 is an intronless gene encoding a member of the SRY-related HMG-box (SOX) family of transcription factors involved in the regulation of embryonic development and in the determination of cell fate. The product of the SOX2 gene is required for stem-cell maintenance in the central nervous system, and also regulates gene expression in the stomach. The SOX2 gene lies within an intron of another gene called SOX2 overlapping transcript (SOX2OT). Further, SOX2 protein may act as a transcriptional activator after forming a protein complex with other proteins. Mutations in the SOX2 gene have been associated with bilateral anophthalmia, a severe form of structural eye malformation, optic nerve hypoplasia and syndromic microphthalmia.
Specifications
| SOX2 | |
| Recombinant Monoclonal | |
| 1.0 mg/mL | |
| Proprietary buffer with 0.008% Bromonitrodioxane, 0.008% Methylisothiazolone; pH 6.8 | |
| P48431, P48432 | |
| SOX2 | |
| Affinity chromatography | |
| RUO | |
| 20674, 6657 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| Btjce | |
| Alexa Fluor Plus 555 | |
| SOX2 | |
| ANOP3; cb236; Delta EF2a; lcc; MCOPS3; MCOPS3 (Microphthalmia Syndromic type 3); RGD1565646; sex determining region Y-box 2; SOX 2; SOX2; Sox-2; soxp; SRY (sex determining region Y) box 2; SRY (sex determining region Y)-box 2; SRY box 2; SRY related HMG box 2; SRY-box 2; SRY-box 2; SOX2; SRY-box containing gene 2; SRY-box transcription factor 2; SRY-related HMG-box gene 2; transcription factor SOX2; transcription factor SOX-2; wu:fb83g04; wu:fc14d07; ysb; zgc:65860; zgc:77389 | |
| Rat | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction