missing translation for 'onlineSavingsMsg'
Learn More

SOX2 Rabbit anti-Human, Mouse, Clone: SOX2/3811R, Novus Biologicals™

Product Code. 18394916 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18394916 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18394916 Supplier Novus Biologicals Supplier No. NBP30789320UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

SOX2 Monoclonal specifically detects SOX2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOX2
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SOX2/3811R
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, ELISA 2-4 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml
Formulation 10 mM PBS with 0.05% BSA
Gene Alias ANOP3, MCOPS3, MGC2413, SRY (sex determining region Y)-box 2, SRY-related HMG-box gene 2, transcription factor SOX2, transcription factor SOX-2
Host Species Rabbit
Immunogen Recombinant fragment (within aa176-305) of human SOX2 protein (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cellular Markers, Core ESC Like Genes, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision
Primary or Secondary Primary
Gene ID (Entrez) 6657
Target Species Human, Mouse
Content And Storage Store at 4°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.