missing translation for 'onlineSavingsMsg'
Learn More

SOX2 Mouse anti-Human, Mouse, Porcine, Clone: 4B9B10, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66411-1-IG-20UL

 View more versions of this product

Product Code. 16838265

  • £135.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is raised against an internal region of human SOX2.

SOX2 is an intronless gene encoding a member of the SRY-related HMG-box (SOX) family of transcription factors involved in the regulation of embryonic development and in the determination of cell fate. The product of the SOX2 gene is required for stem-cell maintece in the central nervous system, and also regulates gene expression in the stomach. The SOX2 gene lies within an intron of another gene called SOX2 overlapping transcript (SOX2OT). Further, SOX2 protein may act as a transcriptional activator after forming a protein complex with other proteins. Mutations in the SOX2 gene have been associated with bilateral anophthalmia, a severe form of structural eye malformation, optic nerve hypoplasia and syndromic microphthalmia.
Specifications

Specifications

SOX2
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P48431, P48432
SOX2
SOX2 Fusion Protein Ag13635
20 μL
Primary
Human, Mouse, Pig
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence
4B9B10
Unconjugated
SOX2
ANOP3, MCOPS3, SOX2, Transcription factor SOX 2
Mouse
Protein G
RUO
20674, 407739, 6657
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.