missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SOX2 Chimeric Recombinant Mouse Monoclonal Antibody (Btjce)

Mouse Recombinant Monoclonal Antibody

£188.00 - £383.00

Specifications

Antigen SOX2 Chimeric
Clone Btjce
Concentration 1.0 mg/mL
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Multiplex Immunohistochemistry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30274830
View Documents
Invitrogen™
740013M
100 μL
£383.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30275367
View Documents
Invitrogen™
740013M20UG
20 μL
£188.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This antibody has Rat Variable heavy and light chains, and Mouse constant domains. It recognizes SOX2 protein. This chimeric antibody is recognized by anti-mouse secondary antibodies. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

SOX2 is an intronless gene encoding a member of the SRY-related HMG-box (SOX) family of transcription factors involved in the regulation of embryonic development and in the determination of cell fate. The product of the SOX2 gene is required for stem-cell maintenance in the central nervous system, and also regulates gene expression in the stomach. The SOX2 gene lies within an intron of another gene called SOX2 overlapping transcript (SOX2OT). Further, SOX2 protein may act as a transcriptional activator after forming a protein complex with other proteins. Mutations in the SOX2 gene have been associated with bilateral anophthalmia, a severe form of structural eye malformation, optic nerve hypoplasia and syndromic microphthalmia.
Specifications

Specifications

SOX2 Chimeric
1.0 mg/mL
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Multiplex Immunohistochemistry
Unconjugated
Mouse
RUO
PBS with 0.09% sodium azide; pH 7.4
P48431, P48432
20674, 499593, 6657
Primary
Affinity chromatography
Btjce
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Recombinant Monoclonal
Liquid
IgG1 κ
Human, Mouse, Rat
SOX2
ANOP3; cb236; Delta EF2a; lcc; MCOPS3; MCOPS3 (Microphthalmia Syndromic type 3); RGD1565646; sex determining region Y-box 2; SOX 2; SOX2; Sox-2; soxp; SRY (sex determining region Y) box 2; SRY (sex determining region Y)-box 2; SRY box 2; SRY related HMG box 2; SRY-box 2; SRY-box 2; SOX2; SRY-box containing gene 2; SRY-box transcription factor 2; SRY-related HMG-box gene 2; transcription factor SOX2; transcription factor SOX-2; wu:fb83g04; wu:fc14d07; ysb; zgc:65860; zgc:77389
SOX2
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.