missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX17 Antibody (614013), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB19241C
This item is not returnable.
View return policy
Description
SOX17 Monoclonal antibody specifically detects SOX17 in Human samples. It is validated for Immunocytochemistry
Specifications
| SOX17 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 64321 | |
| Store at 4C in the dark. | |
| IgG1 |
| Immunocytochemistry | |
| 614013 | |
| Immunocytochemistry | |
| FLJ22252, SRY (sex determining region Y)-box 17, SRY-related HMG-box transcription factor SOX17, transcription factor SOX-17, VUR3 | |
| E. coli-derived recombinant human SOX17, Asp177-Val414, Accession # Q9H6I2 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction