missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX15 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16725-1-AP-20UL
This item is not returnable.
View return policy
Description
SOX15 is a member of the SOX (SRY-related HMG-box) family of transcription factors and is involved in the regulation of embryonic development and in the determination of cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins.Specifications
| SOX15 | |
| Polyclonal | |
| Unconjugated | |
| SOX15 | |
| Protein SOX 12, Protein SOX 15, Protein SOX 20, SOX12, SOX15, SOX20, SOX26, SOX27 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20670, 6665 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O60248, P43267 | |
| SOX15 | |
| SOX15 Fusion Protein Ag10173 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction