missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SOX15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116350
This item is not returnable.
View return policy
Description
SOX15 Polyclonal Antibody for Western Blot
SOX15 is a member of the SOX (SRY-related HMG-box) family of transcription factors and is involved in the regulation of embryonic development and in the determination of cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins.
Specifications
| SOX15 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| O60248 | |
| SOX15 | |
| KLH conjugated synthetic peptide between 152-165 amino acids from the C-terminal region of human (Mouse) Sox15. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| SOX15 | |
| Protein SOX-12; Protein SOX-15; Protein SOX-20; SOX12; SOX15; Sox-15; SOX20; SOX26; SOX27; SRY (sex determining region Y)-box 15; SRY (sex determining region Y)-box 20; SRY box 15; SRY-box 15; SRY-box containing gene 15; SRY-box transcription factor 15 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 6665 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction