missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX14 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£170.00 - £385.00
Specifications
| Antigen | SOX14 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18601401
|
Novus Biologicals
NBP2-94603-0.02ml |
0.02 mL |
£170.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18681111
|
Novus Biologicals
NBP2-94603-0.1ml |
0.1 mL |
£385.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SOX14 Polyclonal antibody specifically detects SOX14 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| SOX14 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| PBS (pH 7.3), 50% glycerol | |
| 8403 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| HMG box transcription factor SOX-14, MGC119898, MGC119899, SRY (sex determining region Y)-box 14, SRY-box 14, transcription factor SOX-14 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 91-240 of human SOX14 (NP_004180.1). RYVFPLPYLGDTDPLKAAGLPVGASDGLLSAPEKARAFLPPASAPYSLLDPAQFSSSAIQKMGEVPHTLATGALPYASTLGYQNGAFGSLSCPSQHTHTHPSPTNPGYVVPCNCTAWSASTLQPPVAYILFPGMTKTGIDPYSSAHATAM | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title