missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX13 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-55489-25ul
This item is not returnable.
View return policy
Description
SOX13 Polyclonal specifically detects SOX13 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| SOX13 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL | |
| Islet cell antigen 12, MGC117216, Sox-13, SRY (sex determining region Y)-box 13ICA12islet cell 12, SRY-box 13, SRY-related HMG-box gene 13, transcription factor SOX-13, Type 1 diabetes autoantigen ICA12 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| SOX13 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:DVLYPRAAGMPLAQPLVEHYVPRSLDPNMPVIVNTCSLREEGEGTDDRHSVADGEMYRYSEDEDSEGEEKSDGELVVLT | |
| 25 μL | |
| Diabetes Research, Lipid and Metabolism | |
| 9580 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction