missing translation for 'onlineSavingsMsg'
Learn More

SOX13 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18414424 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18414424 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18414424 Supplier Novus Biologicals Supplier No. NBP171851JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SOX13 Polyclonal antibody specifically detects SOX13 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOX13
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias Islet cell antigen 12, MGC117216, Sox-13, SRY (sex determining region Y)-box 13ICA12islet cell 12, SRY-box 13, SRY-related HMG-box gene 13, transcription factor SOX-13, Type 1 diabetes autoantigen ICA12
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 572 and 622 of human Sex Determining Region Y-box 13 using the numbering given in entry NP_005677.2 (GeneID 9580).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Diabetes Research, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 9580
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.