missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX13 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-71851F
This item is not returnable.
View return policy
Description
SOX13 Polyclonal antibody specifically detects SOX13 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| SOX13 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| Islet cell antigen 12, MGC117216, Sox-13, SRY (sex determining region Y)-box 13ICA12islet cell 12, SRY-box 13, SRY-related HMG-box gene 13, transcription factor SOX-13, Type 1 diabetes autoantigen ICA12 | |
| The immunogen for this product maps to a region between residue 572 and 622 of human Sex Determining Region Y-box 13 using the numbering given in entry NP_005677.2 (GeneID 9580). | |
| 0.1 mL | |
| Diabetes Research, Lipid and Metabolism | |
| 9580 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| FITC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction