missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX12 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23939-1-AP-150UL
This item is not returnable.
View return policy
Description
Members of the SOX family of transcription factors are characterized by the presence of a DNA-binding high mobility group (HMG) domain, homologous to the HMG box of sex-determining region Y (SRY). Forming a subgroup of the HMG domain superfamily, SOX proteins have been implicated in cell fate decisions in a diverse range of developmental processes. SOX transcription factors have diverse tissue-specific expression patterns during early development and have been proposed to act as target-specific transcription factors and/or as chromatin structure regulatory elements. The protein encoded by this gene was identified as a SOX family member based on conserved domains, and its expression in various tissues suggests a role in both differentiation and maintece of several cell types.Specifications
| SOX12 | |
| Polyclonal | |
| Unconjugated | |
| SOX12 | |
| Protein SOX 22, SOX12, SOX22, Transcription factor SOX 12 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20667, 6666 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O15370, Q04890 | |
| SOX12 | |
| SOX12 Fusion Protein Ag21043 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction