missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SOX12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565921
This item is not returnable.
View return policy
Description
Immunogen sequence: LDCSALDRDPD LQPPSGTSHF EFPD Highest antigen sequence identity to the following orthologs - mouse 96%, rat 96%.
Members of the SOX family of transcription factors are characterized by the presence of a DNA-binding high mobility group (HMG) domain, homologous to the HMG box of sex-determining region Y (SRY). Forming a subgroup of the HMG domain superfamily, SOX proteins have been implicated in cell fate decisions in a diverse range of developmental processes. SOX transcription factors have diverse tissue-specific expression patterns during early development and have been proposed to act as target-specific transcription factors and/or as chromatin structure regulatory elements. The protein encoded by this gene was identified as a SOX family member based on conserved domains, and its expression in various tissues suggests a role in both differentiation and maintenance of several cell types.
Specifications
| SOX12 | |
| Polyclonal | |
| Unconjugated | |
| SOX12 | |
| 2010205A02Rik; Protein SOX-22; SOX12; Sox-12; SOX22; SOX-22 protein; SRY (sex determining region Y)-box 12; SRY box 12; SRY-box 12; SRY-box containing gene 12; SRY-related HMG-box gene 22; transcription factor SOX-12 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6666 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O15370 | |
| SOX12 | |
| Recombinant Human SOX12. Recombinant protein control fragment (Product #RP-106600). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction