missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SOX11 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA547557
This item is not returnable.
View return policy
Description
In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) SOX1, rhSOX2, rhSOX3, rhSOX5, rhSOX6, rhSOX7, rhSOX9, rhSOX10, rhSOX12, rhSOX14, rhSOX15, rhSOX17, and rhSOX21 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
This intronless gene encodes a member of the SOX family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. The protein may function in the developing nervous system and play a role in tumorigenesis.
Specifications
| SOX11 | |
| Polyclonal | |
| Unconjugated | |
| SOX11 | |
| 1110038H03Rik; 6230403H02Rik; AI836553; end1; MRD27; Sox11; Sox-11; SRY; SRY (sex determining region Y)-box 11; SRY (sex-determining region Y)-box 11; SRY-box 11; SRY-box containing gene 11; SRY-box transcription factor 11; SRY-related HMG-box gene 11; transcription factor SOX-11 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 6664 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P35716 | |
| SOX11 | |
| E. coli-derived recombinant human SOX11 Val2-Pro173. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction