Learn More
SOX11 Monoclonal Antibody (SOX11-C1), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 14-9773-95
Description
The MAb SOX11-C1 (C1) recognizes human Sox11. The transcription factor Sox11 is a member of the SOX (sex determining region Y-related HMG (High Mobility Group) Box) family of proteins. Under normal conditions Sox11 is expressed in the developing central nervous system during embryogenesis and has sequence homology to Sox4. Sox11 plays a role in neuronal maturation and epithelial-mesenchymal interactions and is required for the survival of neuronal and mesenchymal progenitor cells. Sox11 is expressed in mantle cell lymphoma (MCL) and in subsets of hairy cell leukemias, Burkitt lymphomas, and B cell lymphoblastic leukemias, but is not expressed in other B cell lymphomas or in normal B lymphocytes. Sox11 is also expressed in epithelial ovarian cancer and gliomas.
This intronless gene encodes a member of the SOX family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. The protein may function in the developing nervous system and play a role in tumorigenesis.
Specifications
| SOX11 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P35716 | |
| SOX11 | |
| Affinity chromatography | |
| RUO | |
| 6664 | |
| 4°C | |
| Liquid |
| ChIP Assay, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| SOX11-C1 | |
| Unconjugated | |
| SOX11 | |
| 1110038H03Rik; 6230403H02Rik; AI836553; end1; MRD27; Sox11; Sox-11; SRY; SRY (sex determining region Y)-box 11; SRY (sex-determining region Y)-box 11; SRY-box 11; SRY-box containing gene 11; SRY-box transcription factor 11; SRY-related HMG-box gene 11; transcription factor SOX-11 | |
| Mouse | |
| 10 mg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.