missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | SOX10 |
|---|---|
| Concentration | 0.4 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16826803
|
Proteintech
10422-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16816803
|
Proteintech
10422-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SOX10 is a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. SOX10 plays a central role in developing and maturing of glia. Specifically, SOX10 activates expression of myelin genes (such as DUSPP15 and MYRF) during oligodendrocyte (OL) maturation; thus, playing a central role in oligodendrocyte maturation and CNS myelination. Once induced MYRF cooperates with SOX10 to implement the myelination program. SOX10 is also a transcriptional activator of MITF acting synergistically with PAX3. Mutations in this gene are associated with Waardenburg-Shah and Waardenburg-Hirschsprung disease.Specifications
| SOX10 | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O55170, P56693, Q04888 | |
| 20665, 29361, 6663 | |
| SOX10 Fusion Protein Ag0673 | |
| Primary | |
| -20°C | |
| SOX10 |
| 0.4 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| DOM, SOX10, Transcription factor SOX 10, WS2E, WS4 | |
| SOX10 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title