missing translation for 'onlineSavingsMsg'
Learn More

SOX10 Polyclonal Antibody, Invitrogen™

Goat Polyclonal Antibody

Brand:  Invitrogen PA537890

Code : NEW

Additional Details : Weight : 0.00900kg

Product Code. 15504831

  • £398.00 / 100µg
Estimated Shipment: 24-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody is predicted to react with mouse, rat, dog, pig and cow based on sequence homology.

SOX10 Polyclonal antibody specifically detects SOX10 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.
Specifications

Specifications

SOX10
Polyclonal
Unconjugated
SOX10
cls; cls/sox10; colorless; colourless; colourless/sox10; DOM; dominant megacolon; dominant megacolon, mouse, human homolog of; golas; gos; HMG-box transcription factor Sox10; MGC15649; PCWH; Protein SOX-21; SOX10; Sox-10; sox10 protein; sox10b; Sox21; SOX9; SRY; SRY (sex determining region Y)-box 10; SRY (sex determining region Y)-box 9 (campomelic dysplasia, autosomal sex-reversal); SRY box 10; SRY-box 10; SRY-box containing gene 10; SRY-box containing protein 10; SRY-box transcription factor 10; SRY-related HMG-box gene 10; transcription factor SOX-10; Transcription factor SOX-M; WS2E; WS4; WS4C; zgc:100757
Goat
Antigen affinity chromatography
RUO
20665, 29361, 6663
-20°C
Liquid
Immunocytochemistry, Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
O55170, P56693, Q04888
SOX10
Peptide with sequence C-DAKAQVKTETAGPQ, from the internal region of SOX10
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers