missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SOX10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112860
This item is not returnable.
View return policy
Description
SOX10 Polyclonal Antibody for Western Blot, ELISA
SOX10 is a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. This protein may act as a transcriptional activator after forming a protein complex with other proteins. It acts as a nucleocytoplasmic shuttle protein and is important for neural crest and peripheral nervous system development. Mutations in this gene are associated with Waardenburg-Shah and Waardenburg-Hirschsprung disease.
Specifications
| SOX10 | |
| Polyclonal | |
| Unconjugated | |
| SOX10 | |
| cls; cls/sox10; colorless; colourless; colourless/sox10; DOM; dominant megacolon; dominant megacolon, mouse, human homolog of; golas; gos; HMG-box transcription factor Sox10; MGC15649; PCWH; Protein SOX-21; SOX10; Sox-10; sox10 protein; sox10b; Sox21; SOX9; SRY; SRY (sex determining region Y)-box 10; SRY (sex determining region Y)-box 9 (campomelic dysplasia, autosomal sex-reversal); SRY box 10; SRY-box 10; SRY-box containing gene 10; SRY-box containing protein 10; SRY-box transcription factor 10; SRY-related HMG-box gene 10; transcription factor SOX-10; Transcription factor SOX-M; WS2E; WS4; WS4C; zgc:100757 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 140616 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| 0 | |
| SOX10 | |
| Recombinant Zebrafish HMG-box transcription factor Sox10 protein (1-485AA). | |
| 100 μg | |
| Primary | |
| Zebrafish | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction