missing translation for 'onlineSavingsMsg'
Learn More

SOX10 Mouse anti-Human, Clone: 1D2C8, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66786-1-IG-150UL

 View more versions of this product

Product Code. 16818985

  • £349.00 / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SOX10 is a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. This protein may act as a transcriptional activator after forming a protein complex with other proteins. It acts as a nucleocytoplasmic shuttle protein and is important for neural crest and peripheral nervous system development. Mutations in this gene are associated with Waardenburg-Shah and Waardenburg-Hirschsprung disease.
Specifications

Specifications

SOX10
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P56693
SOX10
SOX10 Fusion Protein Ag23191
150 μL
Primary
Human
Antibody
IgG2a
Immunohistochemistry (Paraffin), Western Blot
1D2C8
Unconjugated
SOX10
DOM, SOX10, Transcription factor SOX 10, WS2E, WS4
Mouse
Protein A
RUO
6663
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.