missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX10 Antibody (CL4455), Novus Biologicals™
Mouse Monoclonal Antibody
£272.00 - £460.00
Specifications
| Antigen | SOX10 |
|---|---|
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Host Species | Mouse |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18093466
|
Novus Biologicals
NBP2-59050 |
100 μL |
£460.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18693338
|
Novus Biologicals
NBP2-59050-25ul |
25 μL |
£272.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SOX10 Monoclonal specifically detects SOX10 in Human, Mouse, Feline samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| SOX10 | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| SOX10 | |
| IgG1 | |
| Protein A purified |
| Monoclonal | |
| Purified | |
| RUO | |
| 6663 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:PHYTDQPSTSQIAYTSLSLPHYGSAFPSISRPQFDYSDHQPSGPYYGHSG | |
| Primary | |
| Specificity of human SOX10 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title