missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX1 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
£161.00 - £383.00
Specifications
| Antigen | SOX1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA Recommended starting concentration is 1 μg/mL |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30232300
|
Novus Biologicals
NBP3-33424-20ul |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30231687
|
Novus Biologicals
NBP3-33424-100ul |
100 μL |
£383.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SOX1 Monoclonal antibody specifically detects SOX1 in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| SOX1 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Stem Cells | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| 6656 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA Recommended starting concentration is 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| SRY (sex determining region Y)-box 1, SRY-related HMG-box gene 1, transcription factor SOX-1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human SOX1 (NP_005977.2).,, Sequence:, MYSMMMETDLHSPGGAQAPTNLSGPAGAGGGGGGGGGGGGGGGAKANQDRVKRPMNAFMVWSRGQRRKMAQENPKMHNSEISKRLGAEWKVMSEAEKRPF | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title