missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX1 Antibody [mFluor Violet 610 SE], Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals AF3369MFV610
This item is not returnable.
View return policy
Description
SOX1 Polyclonal antibody specifically detects SOX1 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry
Specifications
| SOX1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Goat | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| mFluor Violet 610 SE | |
| SRY (sex determining region Y)-box 1, SRY-related HMG-box gene 1, transcription factor SOX-1 | |
| E. coli-derived recombinant human SOX1, Asn242-Gly379 (Leu276Ile), Accession # NP_005977 | |
| 0.1 mL | |
| Stem Cells | |
| 6656 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction