missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SOX1 Polyclonal antibody specifically detects SOX1 in Human, Mouse, Equine, Opossum, Primate, Xenopus samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | SOX1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | APC |
| Dilution | Western Blot |
| Formulation | PBS |
| Gene Alias | SRY (sex determining region Y)-box 1, SRY-related HMG-box gene 1, transcription factor SOX-1 |
| Host Species | Rabbit |
| Immunogen | A portion of amino acids 100-150 of human SOX1 was used as the immunogen. |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?