missing translation for 'onlineSavingsMsg'
Learn More

Cytiva SOURCE™ 15S Chromatography Media

Product Code. 11520804 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
Unit Size:
50mL
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11520804 10 mL Pack of 10
11510804 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11520804 Supplier Cytiva Supplier No. 17094410

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemically stable cation exchanger for high resolution polishing purification of proteins, peptides and oligonucleotides

  • Average particle size: 15μm
  • Ligand: sulphonate group
  • Matrix: polystyrene/divinylbenzene
  • pH stability working range: 2 to 13
  • Available in 10mL and 50mL bottles

TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mL
Stability Stable in common Ion Exchange Buffers
For Use With (Application) For High Resolution Polishing PurifiCatIon of Proteins, Peptides and Oligonucleotides
Storage Requirements 4°C to 30°C
Product Type Source 15 S Media
Flow Rate 400 cm/Hoμr
Particle Size 15 μm
Height (English) Bed 3.93 in.
Height (Metric) Bed 10 cm
Ion Exchange Type Strong CatIon Exchanger
Max. Pressure 100 kPa
Matrix Polystyrene/Divinylbenzene
pH Range 1 to 14
Storage Buffer 20% Ethanol, 0.2 M Sodium Acetate
Ligand Type Sulphonate Group
Show More Show Less
compliance-icons
Product Identifier
  • SOURCE 15S, 10 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
  • UNKNOWN ACUTE-18 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.