missing translation for 'onlineSavingsMsg'
Learn More

Sotrastaurin, MedChemExpress

Product Code. 16433848
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16433848

Brand: Medchem Express HY10343

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sotrastaurin (AEB071) is a potent and orally-active pan-PKC inhibitor, with Kis of 0.22 nM, 0.64 nM, 0.95 nM, 1.8 nM, 2.1 nM and 3.2 nM for PKCθ, PKCβ, PKCα, PKCη, PKCδ and PKCε, respectively.

Sotrastaurin (AEB071) is a potent and orally-active pan-PKC inhibitor, with Kis of 0.22 nM, 0.64 nM, 0.95 nM, 1.8 nM, 2.1 nM and 3.2 nM for PKCθ, PKCβ, PKCα, PKCη, PKCδ and PKCε, respectively.

Specifications

Chemical Name or Material Sotrastaurin
CAS 425637-18-9
Color Red
Purity Grade Notes Research
Molecular Formula C25H22N6O2
Quantity 10 mg
Synonym AEB071
Solubility Information DMSO : ≥ 50 mg/mL (114.03 mM)
SMILES O=C(C(C1=C2C=CC=CC2=NC(N3CCN(C)CC3)=N1)=C4C5=CNC6=C5C=CC=C6)NC4=O
Molecular Weight (g/mol) 438.48
Formula Weight 438.48
Percent Purity 99.89%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.