missing translation for 'onlineSavingsMsg'
Learn More

Sotrastaurin, MedChemExpress

Product Code. 16486567
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16486567 5 mg 5mg
16460217 50 mg 50mg
16433848 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16486567 Supplier Medchem Express Supplier No. HY10343
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Sotrastaurin (AEB071) is a potent and orally-active pan-PKC inhibitor, with Kis of 0.22 nM, 0.64 nM, 0.95 nM, 1.8 nM, 2.1 nM and 3.2 nM for PKCθ, PKCβ, PKCα, PKCη, PKCδ and PKCε, respectively.

Sotrastaurin (AEB071) is a potent and orally-active pan-PKC inhibitor, with Kis of 0.22 nM, 0.64 nM, 0.95 nM, 1.8 nM, 2.1 nM and 3.2 nM for PKCθ, PKCβ, PKCα, PKCη, PKCδ and PKCε, respectively.

Specifications

Chemical Name or Material Sotrastaurin
CAS 425637-18-9
Color Red
Purity Grade Notes Research
Molecular Formula C25H22N6O2
Quantity 5 mg
Synonym AEB071
Solubility Information DMSO : ≥ 50 mg/mL (114.03 mM)
SMILES O=C(C(C1=C2C=CC=CC2=NC(N3CCN(C)CC3)=N1)=C4C5=CNC6=C5C=CC=C6)NC4=O
Molecular Weight (g/mol) 438.48
Formula Weight 438.48
Percent Purity 99.89%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.