missing translation for 'onlineSavingsMsg'
Learn More

Sotorasib, MedChemExpress

Product Code. 16491067
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491067 50 mg 50mg
16454808 10 mg 10mg
16435606 100 mg 100mg
16427557 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16491067 Supplier Medchem Express Supplier No. HY114277

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sotorasib (AMG-510) is a first-in-class, orally bioavailable, and selective KRAS G12C covalent inhibitor. Sotorasib irreversibly inhibits KRAS G12C by locking it in an inactive GDP-bound state. Sotorasib is the first KRAS G12C inhibitor in clinical development and leads to the regression of KRAS G12C tumors.

Sotorasib (AMG-510) is a first-in-class, orally bioavailable, and selective KRAS G12C covalent inhibitor. Sotorasib irreversibly inhibits KRAS G12C by locking it in an inactive GDP-bound state. Sotorasib is the first KRAS G12C inhibitor in clinical development and leads to the regression of KRAS G12C tumors.

Specifications

Chemical Name or Material Sotorasib
Physical Form Powder
CAS 2296729-00-3
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C30H30F2N6O3
For Use With (Application) Cancer-Kinase/protease
Synonym AMG-510
Solubility Information DMSO : 50 mg/mL (89.19 mM; Need ultrasonic) ∣H2O : 33.33 mg/mL (59.46 mM; ultrasonic and adjust pH to 11 with NaOH)
SMILES O=C(C=C)N1C[C@H](C)N(C2=NC(N(C3=C(C)C=CN=C3C(C)C)C4=C2C=C(F)C(C5=C(O)C=CC=C5F)=N4)=O)CC1
Molecular Weight (g/mol) 560.59
Formula Weight 560.59
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.