missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Sostdc1 Monoclonal Antibody (C1), Invitrogen™
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA543568

 View more versions of this product

Product Code. 17644026

  • £386.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

May be involved in the onset of endometrial receptivity for implantation/sensitization for the decidual cell reaction Enhances Wnt signaling and inhibits TGF-beta signaling. Directly antagonizes activity of BMP2, BMP4, BMP6 and BMP7 in a dose-dependent manner.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sostdc1
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Q6X4U4
SOSTDC1
Phe24-Ser206 (Accession Q6X4U4), with N-terminal His Tag.
100 μL
Primary
Human
Antibody
IgG1 κ
Western Blot
C1
Unconjugated
SOSTDC1
0610006G05Rik; CDA019; cystine-knot containing secreted protein; DAND7; Ectodermal BMP inhibitor; ECTODIN; sclerostin domain containing 1; sclerostin domain-containing protein 1; Sclerostin-like protein; similar to cystine knot-containing secreted protein; Sostdc1; Sostl; USAG1; USAG-1; Uterine sensitization-associated gene 1 protein; uterine sensitization-associated protein 1; uterine sensitization-associated protein-1; Wise; Wnt-signaling modulator
Mouse
Protein A/G
RUO
25928
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.