missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Sostdc1 Monoclonal Antibody (C1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA543569
This item is not returnable.
View return policy
Description
May be involved in the onset of endometrial receptivity for implantation/sensitization for the decidual cell reaction Enhances Wnt signaling and inhibits TGF-beta signaling. Directly antagonizes activity of BMP2, BMP4, BMP6 and BMP7 in a dose-dependent manner.
Specifications
| Sostdc1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q6X4U4 | |
| SOSTDC1 | |
| Phe24-Ser206 (Accession Q6X4U4), with N-terminal His Tag. | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Western Blot | |
| C1 | |
| Unconjugated | |
| SOSTDC1 | |
| 0610006G05Rik; CDA019; cystine-knot containing secreted protein; DAND7; Ectodermal BMP inhibitor; ECTODIN; sclerostin domain containing 1; sclerostin domain-containing protein 1; Sclerostin-like protein; similar to cystine knot-containing secreted protein; Sostdc1; Sostl; USAG1; USAG-1; Uterine sensitization-associated gene 1 protein; uterine sensitization-associated protein 1; uterine sensitization-associated protein-1; Wise; Wnt-signaling modulator | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 25928 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction