missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOST/Sclerostin Antibody (RM0129-4D77), Alexa Fluor™ 488, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-22506AF488
This item is not returnable.
View return policy
Description
SOST/Sclerostin Monoclonal specifically detects SOST/Sclerostin in Mouse samples. It is validated for Western Blot.
Specifications
| SOST/Sclerostin | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| sclerostin, VBCHsclerosteosis | |
| Rat | |
| Protein A or G purified | |
| Signal Transduction | |
| 50964 | |
| Store at 4C in the dark. | |
| IgG2 |
| Western Blot | |
| RM0129-4D77 | |
| Western Blot | |
| Q99P68 | |
| SOST | |
| Mouse recombinant protein of Sclerostin | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction