missing translation for 'onlineSavingsMsg'
Learn More

SOS2 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP1-05958DL594

Product Code. 18440407

  • £520.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SOS2 Polyclonal antibody specifically detects SOS2 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SOS2
Polyclonal
Western Blot
FLJ25596, guanine nucleotide exchange factor, guanine nucleotide releasing factor, son of sevenless (Drosophilia) homolog 2, son of sevenless homolog 2, son of sevenless homolog 2 (Drosophila), SOS-2
The immunogen recognized by this antibody maps to a region between residue 1282 and 1332 of human son of sevenless ortholog 2 using the numbering given in entry NP_008870.2 (GeneID 6655).
0.1 mL
Breast Cancer, Cancer
6655
Store at 4°C in the dark.
IgG
Western Blot
DyLight 594
50mM Sodium Borate
Rabbit
Immunogen affinity purified
RUO
Primary
Human, Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.