missing translation for 'onlineSavingsMsg'
Learn More

SOS2 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18432405 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18432405 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18432405 Supplier Novus Biologicals Supplier No. NBP105958AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SOS2 Polyclonal antibody specifically detects SOS2 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOS2
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias FLJ25596, guanine nucleotide exchange factor, guanine nucleotide releasing factor, son of sevenless (Drosophilia) homolog 2, son of sevenless homolog 2, son of sevenless homolog 2 (Drosophila), SOS-2
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1282 and 1332 of human son of sevenless ortholog 2 using the numbering given in entry NP_008870.2 (GeneID 6655).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 6655
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.